Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MS-VJP-M03-01-ST-N MS-VKP-M13-01-ST-N MS-VJP-M05-01-ST-N MS-VJP-M06-01-ST-N MS-VJP-M04-01-ST-N MS-VJP-M02-01-ST-N MS-VKP-M09-01-ST-N MS-VKP-M07-01-ST-N MS-VJP-M01-01-ST-N MS-VLP-M16-01-ST-N MS-VKP-M08-01-ST-N MS-VKP-M11-01-ST-N MS-VKP-M14-01-ST-N MS-VKP-M12-01-ST-N
MFLX32447-50EA 1440.96 CAD
MFLX32447-50 MFLX32447-60 MFLX32447-30 MFLX32447-52 MFLX32447-62 MFLX32447-40 MFLX32447-10 MFLX32447-54 MFLX32447-32 MFLX32447-20 MFLX32447-42 MFLX32447-34 MFLX32447-00 MFLX32447-02
Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Flow Meters
Ideal for the most aggressive process applications.
  • Thick polycarbonate front shield provides protection
  • Unique design facilitates ease of maintenance
  • Easy-to-read direct-reading scale offers both metric and English measurements
  • Available in fluted or plain tapered tubes
Detachable, clear 3/16"-thick polycarbonate front shield provides protection at maximum rated temperature and pressure. Easily read the graduated direct-reading scale. The interchangeable transparent scale plates mount directly on the front of the flowtube. Unique float stops allow easy cleaning-remove and replace the float without disturbing either the flowtube or the shield. Tube sizes 3, 4, and 5 are fluted; tube sizes 6, 8, and 9 are tapered with a stainless steel guide rod.