Masterflex® Thermal Mass Flowmeters/Flow Transmitters for Gases, Avantor®
Supplier: VWR International
50D-4 50D-5 50D-6 50D-8 50D-13-S7 MFLX32707-00 MFLX32707-02 MFLX32707-04 MFLX32707-06 MFLX32707-08 MFLX32707-10 MFLX32707-12 MFLX32707-16 MFLX32707-34 MFLX32711-40 MFLX32712-40 MFLX32712-52 MFLX32712-36
MFLX32707-14EA 805.8 CAD
MFLX32707-14 MFLX32707-20 MFLX32707-22 MFLX32707-24 MFLX32707-26 MFLX32707-28 MFLX32707-30 MFLX32707-32 MFLX32707-36 MFLX32711-36 MFLX32712-44 MFLX32707-00 MFLX32707-02 MFLX32707-04 MFLX32707-06 MFLX32707-08 MFLX32707-10 MFLX32707-12 MFLX32707-16 MFLX32707-34 MFLX32711-40 MFLX32712-40 MFLX32712-52 MFLX32712-36
Masterflex® Thermal Mass Flowmeters/Flow Transmitters for Gases, Avantor®
Flow Meters
Get faster performance with miniature coil design.
  • All meters feature 0 to 5 VDC output signal for data logging or controlling other instruments
These low-cost flowmeters use thermal gas flow sensing technique that results in highly accurate readings and repeatability. All units are calibrated to air/nitrogen—call our Application Specialists for meters calibrated to other gases or gas mixtures. Meters feature a cable hub connector for a quick connection to a power supply and 0 to 5 VDC output cable; order power supplies/cables under accessories.
Flowmeters have low pressure drop across the sensor. The output signal can connect to a remote display, recorder, or any instrument that accepts a 0 to 5 VDC signal. Compact size makes it easy to carry around.
Wetted materials are anodized aluminum, 316 stainless steel, Viton® O-rings, and 316 stainless steel, and acetal fittings. Order rechargeable battery kit under Accessories.
Delivery information: Includes two compression fittings and 36" (0.9 m) cable with bub connector.