Masterflex® Direct Reading Variable Area Flowmeters for Water, PFA Housing, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
L53/B1 L53/B3 L54/A2 L54/A5 L54/C1 MFLX32050-00 MFLX32050-02 MFLX32050-04 MFLX32050-08 MFLX32050-12 MFLX32050-16 MFLX32051-00 MFLX32051-10 MFLX32051-16
MFLX32050-10EA 1396.72 CAD
MFLX32050-10 MFLX32050-14 MFLX32051-02 MFLX32051-08 MFLX32051-18 MFLX32050-00 MFLX32050-02 MFLX32050-04 MFLX32050-08 MFLX32050-12 MFLX32050-16 MFLX32051-00 MFLX32051-10 MFLX32051-16
Masterflex® Direct Reading Variable Area Flowmeters for Water, PFA Housing, Avantor®
Flow Meters
A unique combination of rugged frame and chemically inert wetted materials.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with water for laboratory and industrial applications
  • Rigid aluminum frame is PFA coated to resist ambient corrosives
  • Ideal for applications involving ultrapure water or highly corrosive solvents
These flowmeters are great for use with etchants and high purity water like those used in the semiconductor industry. Each flowmeter is individually tested on aMore Product Information