Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 150-mm, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
P04/1-N044-40G P04/1-N044-40ST P04/1-N044-40C T54/1-N044-40S P24/1-N044-40G P24/1-N044-40ST P24/1-N044-40C P14/1-N044-40ST P14/1-N044-40C T54/1-044-40G P14/1-N044-40G
MFLX03217-32EA 374.46 CAD
MFLX03217-32 MFLX03217-34 MFLX03217-36 MFLX03217-90 MFLX03219-33 MFLX03219-35 MFLX03219-37 MFLX03294-34 MFLX03294-36 MFLX03217-87 MFLX03294-32
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 150-mm, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet,More Product Information