Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
TPG MFLX03226-10 MFLX03226-30 TP3 TP2 MFLX32462-50 MFLX32462-55
MFLX03218-58EA 102.7 CAD
MFLX03218-58 MFLX03226-10 MFLX03226-30 MFLX03226-40 MFLX03226-50 MFLX32462-50 MFLX32462-55
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Flow Meters
Easily mount flowmeters on tripod for easy viewing.
  • Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.