Gast Direct-Drive Foot-Mount Air Motors
Supplier: GAST MANUFACTURING CORP
8AM-FRV-2B MFLX07055-50
MFLX07055-82EA 919.56 CAD
MFLX07055-82 MFLX07055-50
Gast Direct-Drive Foot-Mount Air Motors
Pumps Motors
Four-vane rotary design is almost maintenance free.
  • Nonsparking, nonelectrical operation—safe and reliable
These combination filter/regulators save space and set-up time. Each has a manual drain and polycarbonate bowl. Temperature range is 0 to 125 °F (-18 to 51 °C). A 0 to 160 psi gauge is standard except on model 98252-00. Order gauge 68002-05.
Delivery information: Includes motors come with an exhaust muffler. Install a compressed-air filter in-line to remove oil, water, and solid contaminants—order connecting nipples and filters separately.