Masterflex® L/S® Precision Washdown Modular Drives with Wall-Mount Controller, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
7557-60 7557-60 7557-70 7557-70 7557-70 7557-70 7557-70 7557-60 7557-70 7557-60 7557-60 7557-60 7557-70 7557-60 7557-70 7557-70 7557-60 7557-60
MFLX07557-60-ITCS 0 CAD
MFLX07557-60-IT MFLX07557-70-UK MFLX07557-60-DK MFLX07557-70-EU MFLX07557-60-IL MFLX07557-60-IN MFLX07557-60-CH MFLX07557-70-AU MFLX07557-70-DK MFLX07557-60 MFLX07557-70 MFLX07557-70-IT MFLX07557-60-AU MFLX07557-60-UK MFLX07557-70-CH MFLX07557-70-IN MFLX07557-60-EU MFLX07557-70-IL
Masterflex® L/S® Precision Washdown Modular Drives with Wall-Mount Controller, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
  • Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm and tubing size
  • Three digit LED display shows motor rpm — confirm speed setting at a glance
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Wall-mount controller housing is powder-coated aluminum, IP66/NEMA 4X rated for washdown
  • Remote control capabilities via weather-resistant connection on controller
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Delivery information: Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.