Masterflex® L/S® Precision Modular Drives with Benchtop Controller, Avantor®
Supplier: Avantor Fluid Handling
7557-00 7557-10 7557-00 7557-10 7557-00 7557-10 7557-00 7557-10 7557-00 7557-10 7557-00 7557-10 7557-00 7557-10 7557-00 7557-10 7557-00 7557-10
MFLX07557-00CS 0 CAD
MFLX07557-00 MFLX07557-10 MFLX07557-00-AU MFLX07557-10-AU MFLX07557-00-DK MFLX07557-10-DK MFLX07557-00-IN MFLX07557-10-IN MFLX07557-00-IL MFLX07557-10-IL MFLX07557-00-EU MFLX07557-10-EU MFLX07557-00-UK MFLX07557-10-UK MFLX07557-00-CH MFLX07557-10-CH MFLX07557-00-IT MFLX07557-10-IT
Masterflex® L/S® Precision Modular Drives with Benchtop Controller, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
  • Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm and tubing size
  • Three digit LED display shows motor rpm — confirm speed setting at a glance
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Splash-resistant ABS plastic controller housing is IP33
  • rated
  • Remote control capabilities via a DB9 female connector on back of controller
L/S® modular drives feature reversibleMore Product Information