Masterflex® L/S® Compact Variable-Speed Drive, Avantor®
Supplier: Avantor Fluid Handling
77200-12 77200-20 77200-20 77200-20 77200-20 77200-20 77200-20 77200-20 77200-20 77200-20
MFLX77200-12EA 1519.96 CAD
MFLX77200-12 MFLX77200-20 MFLX77200-20-AU MFLX77200-20-DK MFLX77200-20-IN MFLX77200-20-IL MFLX77200-20-EU MFLX77200-20-UK MFLX77200-20-CH MFLX77200-20-IT
Masterflex® L/S® Compact Variable-Speed Drive, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Stackable, compact pump drive delivers versatile performance in limited space.
  • Flow rate 2.1 to 560 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Stackable, compact drive is perfect for small work areas
  • Constructed with a painted steel housing with IP22 rating
  • No difficult programming—single switch for forward/off/reverse and analog controller
  • Pump in either direction—also allows purging before or after pumping
  • Remote control of start/stop via contact closure
These compact drives are a perfect when space is limited, as they fitMore Product Information