Masterflex® L/S® Easy-Load® II Pump Heads with 2-Channels for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
77202-50 77202-60
MFLX77202-50EA 935.36 CAD
MFLX77202-50 MFLX77202-60
Masterflex® L/S® Easy-Load® II Pump Heads with 2-Channels for Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
No need to stack heads—get two channels in one head.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Compact design for efficient use of space
  • Suitable for use with L/S® Precision pump tubing sizes
The two-channel pump head lets you pump two synchronous flow channels, proper retention requires the use of the same size tubing in each channel. These pump heads accept the four L/S® precision tubing sizes L/S® 13, L/S® 14, L/S® 16, and L/S® 25. Achieve flow rates of 0.06 to 1000 ml/min—flow rate depends on drive rpm and tubing size.
Delivery information: Includes mounting hardware.