Masterflex® L/S® PTFE-Diaphragm Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
7090-42 7090-62
MFLX07090-42EA 1722.2 CAD
MFLX07090-42 MFLX07090-62
Masterflex® L/S® PTFE-Diaphragm Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Compatible with most Masterflex® L/S® drives.
  • PTFE and glass wetted parts help maintain fluid purity
  • Ideal for pumping through filters or into pressurized systems
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
Note: Do not use these pump heads with Masterflex® drives requiring a long-shaft pump head.
Delivery information: Includes mounting tools, coupling hardware, and service kit.