Masterflex® L/S® Easy-Load® II Pump Heads for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
77200-30 77200-50 77200-60 77201-60
MFLX77200-30CS 0 CAD
MFLX77200-30 MFLX77200-50 MFLX77200-60 MFLX77201-60
Masterflex® L/S® Easy-Load® II Pump Heads for Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Model available with integrated open-head sensor–connects directly to sensor-equipped drives
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S®
More Product Information