Masterflex® Multi-Syringe Pumps, Multi-Step, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
74900-85 74901-15 74900-85 74901-15 74901-05 74901-05 780222 74901-15 789202 78-9232C 780212 74901-05 74900-65 74901-05 74900-65 74900-65 74900-85 74900-65 74900-85 74901-05 789222 74900-85 74901-05 74901-05 74900-65 74901-15 789212C 74901-05 74900-85 780202 74901-15 74900-65 74900-85 74901-15 74900-65 74900-85 74900-65 74901-15 74901-15 74900-85 74900-65
MFLX74901-05EA 9140.3 CAD
MFLX74900-85-DK MFLX74900-65-IT MFLX74900-65-AU DUP-DO-NOT-USE080 MFLX74900-65-EU MFLX74901-15-UK MFLX74901-05-DK MFLX74900-85-UK DUP-DO-NOT-USE085 MFLX74901-05-AU MFLX74901-15-IL MFLX74901-05 MFLX74901-05-EU MFLX74900-65-DK MFLX74901-15-IT MFLX74901-15-IN MFLX74900-85-IL MFLX74901-15-CH MFLX74900-85-CH MFLX74900-85-IN MFLX74901-15-EU MFLX74901-05-IL MFLX74900-85-IT MFLX74900-85-EU MFLX74900-65-UK MFLX74901-05-IT MFLX74901-15-AU MFLX74901-05-IN MFLX74901-05-CH MFLX74900-85-AU DUP-DO-NOT-USE010 DUP-DO-NOT-USE490 DUP-DO-NOT-USE491 MFLX74901-15 MFLX74900-65-CH MFLX74900-65-IN MFLX74900-65 MFLX74900-85 MFLX74901-05-UK MFLX74900-65-IL MFLX74901-15-DK
Masterflex® Multi-Syringe Pumps, Multi-Step, Avantor®
Pumps Syringe Pumps
Ideal for low-flow, low-pressure applications where precision is needed.
  • Flow ranges from 0.05 uL/min up to 70.56 ml/min
  • Built-in program lets you enter up to eight commands to perform a variety of functions—you can even repeat commands up to ten times; all settings are stored in nonvolatile memory
  • Backlit alphanumeric LCD ste
Program flow rate hold, pause, and change of direction; each command can be set to run for up to 12 hours. You can even program infusion/withdrawal pumps to sequence operations: either withdrawal followed by infusion or infusion followed by withdrawal. Ch
Delivery information: Includes a 6-ft cord with grounded plug (U.S. standard plug on 115 VAC model and European plug on 230 VAC models).