Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
78018-20 78018-14 78018-12 78018-10 78018-20 78018-10 78018-12 78018-24 78018-14 78018-10 78018-14 78018-22 78018-20 78018-22 78018-22 78018-14 78018-10 78018-22 78018-24 78018-22 78018-12 78018-22 78018-20 78018-12 78018-10 78018-14 78018-12 78018-12 78018-22 78018-20 78018-12 78018-20 78018-12 78018-24 78018-14 78018-24 78018-14 78018-10 78018-24 78018-24 78018-14 78018-24 78018-10 78018-12 78018-14 78018-22 78018-10 78018-24 78018-10 78018-20 78018-24 78018-22 78018-20 78018-20
MFLX78018-20-DKCS 0 CAD
MFLX78018-20-DK MFLX78018-12-AU MFLX78018-24-DK MFLX78018-12-EU MFLX78018-10-DK MFLX78018-22-EU MFLX78018-14-DK MFLX78018-22-AU MFLX78018-20-CH MFLX78018-22-IT MFLX78018-24-CH MFLX78018-12-UK MFLX78018-22-IN MFLX78018-22-IL MFLX78018-12-IT MFLX78018-12-IN MFLX78018-10-CH MFLX78018-22-UK MFLX78018-14-CH MFLX78018-12-IL MFLX78018-24 MFLX78018-22 MFLX78018-22-DK MFLX78018-20 MFLX78018-10-AU MFLX78018-14-AU MFLX78018-10-EU MFLX78018-20-EU MFLX78018-12-DK MFLX78018-20-AU MFLX78018-24-AU MFLX78018-14 MFLX78018-20-IN MFLX78018-12 MFLX78018-24-EU MFLX78018-10-UK MFLX78018-20-IT MFLX78018-10 MFLX78018-24-IT MFLX78018-24-IN MFLX78018-14-UK MFLX78018-20-IL MFLX78018-22-CH MFLX78018-24-IL MFLX78018-14-EU MFLX78018-10-IT MFLX78018-20-UK MFLX78018-14-IT MFLX78018-12-CH MFLX78018-24-UK MFLX78018-10-IL MFLX78018-10-IN MFLX78018-14-IN MFLX78018-14-IL
Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
Pumps Peristaltic Pumps
Versatile multichannel pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Anti-drip and speed ramping available in Volume dispense mode – ensure accuracy and minimize sample loss
  • Cassette pump models accept 3-stop tubing for easy loading and secure retention
  • Three-level user access – gives you the option of controlling who can operate the pump and at what level
MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters – including speed, flow rate, dispense volume, and more. You can monitor critical processes running 24/7 without the need for onsite personnel while push notifications alert you to operating conditions and error messages.
Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive; USB connection allows for downloadable firmware updates to be loaded into the pump.
Tubing cassettes (included) accept 3-stop tubing and are easily snapped in/out for quick tubing changes. Accepted tubing ranges from 0.13 to 3.17 mm ID for a wide flow range.
Order the three-stop tubing size and formulation to suit your application needs (sold separately).
Ordering information: Each model includes a country specific plug/cord set; choose the catalog number with desired country plug type. All units include an IEC 320 socket with a detachable standard US three prong power cable. See country plug description https://us.vwr.com/cms/tech-article-ac-power-plugs-sockets-universal-pattern-guide.
Delivery information: Includes pump drive, pump head, cassettes, and power adapter with 1.8 m (6 ft.) line cord. These models are shipped with country specific plug/cord set; please order the specific catalog number with the desired plug type.