Masterflex® L/S® Precision Variable-Speed Modular Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
77913-60 77913-70 77913-60 77913-70 77913-60 77913-70 77913-60 77913-70 77913-60 77913-70 77913-60 77913-70 77913-60 77913-70 77913-60 77913-70 77913-60 77913-70 77913-80 77913-80 77913-80 77913-80 77913-80 77913-80 77913-80 77913-80 77913-80
MFLX77913-60EA 4518.8 CAD
MFLX77913-60 MFLX77913-70 MFLX77913-60-AU MFLX77913-70-AU MFLX77913-60-DK MFLX77913-70-DK MFLX77913-60-IN MFLX77913-70-IN MFLX77913-60-IL MFLX77913-70-IL MFLX77913-60-EU MFLX77913-70-EU MFLX77913-60-UK MFLX77913-70-UK MFLX77913-60-CH MFLX77913-70-CH MFLX77913-60-IT MFLX77913-70-IT MFLX77913-80 MFLX77913-80-AU MFLX77913-80-DK MFLX77913-80-IN MFLX77913-80-IL MFLX77913-80-EU MFLX77913-80-UK MFLX77913-80-CH MFLX77913-80-IT
Masterflex® L/S® Precision Variable-Speed Modular Pump Systems, Avantor®
Pumps Peristaltic Pumps
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
  • Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm and tubing size
  • Three digit LED display shows motor rpm — confirm speed setting at a glance
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Choose between benchtop controller or washdown wall-mount controller drives
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Benchtop controller drives feature a splash-resistant ABS plastic controller housing with IP33 rating and remote control capabilities via a DB9 female connector on back of controller. Wall-mount controller housing is powder-coated aluminum and IP66/NEMA 4X rated for washdown with remote control capabilities via weather-resistant connection on controller.