Masterflex® L/S® Replacement Brushes and Brush Caps, Avantor®
Supplier: Avantor
7520-04 7520-06 7533-01
MFLX07520-03EA 73.15 CAD
MFLX07520-03 MFLX07520-04 MFLX07520-06 MFLX07533-01
Masterflex® L/S® Replacement Brushes and Brush Caps, Avantor®
Pumps Peristaltic Pumps
Quickly and easily replace components of your Masterflex system.
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.