Masterflex® Complete Drum and Container Pumps, Avantor®
Supplier: VWR International
MFLX70613-00 MFLX70613-10 MFLX70613-15 MFLX70613-20 MFLX70613-30 MFLX70613-40 MFLX70613-45 MFLX70613-50 MFLX70613-05 MFLX70613-25 MFLX70613-35 MFLX70613-55
MFLX70613-00EA 2512.2 CAD
MFLX70613-00 MFLX70613-10 MFLX70613-15 MFLX70613-20 MFLX70613-30 MFLX70613-40 MFLX70613-45 MFLX70613-50 MFLX70613-05 MFLX70613-25 MFLX70613-35 MFLX70613-55
Masterflex® Complete Drum and Container Pumps, Avantor®
Pumps Drum/Transfer Pumps
System includes pump tube and motor—tube material options ensure chemical compatibility.
  • Flow rate 35 GMP for all models
  • Equipped with variable-speed control motors (except air and explosion-proof models) for added safety and control
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Materials such as PP, PVDF, CPVC, and 316 SS provide excellent chemical resistance against corrosive liquids. Order optional dispensing nozzle from "Accessories" for added control.
Delivery information: Includes electric motor with 16-ft power cord and pump tube.