Masterflex® Chemically Inert Metering Valves, Avantor®
Supplier: VWR International
MVT3 MVT6 VTF-TT-0A
MFLX06394-00EA 189.6 CAD
MFLX06394-00 MFLX06394-02 MFLX06394-04 MFLX06394-16
Masterflex® Chemically Inert Metering Valves, Avantor®
Valves Metering Valves
Easily meter corrosive fluids or gases.
  • Just turn—eight turns fully open or close valv
Excellent for metering corrosive fluids and gases — all wetted parts are CTFE and PTFE. Eight turns will fully open or close valves. Maximum pressure is 75 psi and maximum temperature is 150 °F (93 °C); orifice size is 1/4".