Masterflex® Plastic Ball Valves, Three-way, PVC, Avantor®
Supplier: SPECIALTY MFG CO
PVC326-2F2F2F-E PVC326-2F2F2F-F PVC-326-4F4F4F-F PVC-326-4B4B4B-E PVC-326-4B4B4B-F PVC-326-4SMC4SMC4SMC-E PVC-326-4CJG4CJG4CJG-F PVC-326-6B6B6B-E PVC-326-6B6B6B-F PVC-326-4F4F4F-E PVC-326-4B4B4B-B PVC-326-6B6B6B-B PVC-326-8B8B8B-E PVC-326-8B8B8B-F MFLX01377-76 MFLX01377-78 MFLX01378-97 MFLX01378-98 MFLX01379-05 MFLX01379-02
MFLX01377-58EA 53.25 CAD
MFLX01377-58 MFLX01377-60 MFLX01377-62 MFLX01377-64 MFLX01377-66 MFLX01377-68 MFLX01377-70 MFLX01377-72 MFLX01377-74 MFLX01378-99 MFLX01379-01 MFLX01379-03 MFLX01379-06 MFLX01379-07 MFLX01377-76 MFLX01377-78 MFLX01378-97 MFLX01378-98 MFLX01379-05 MFLX01379-02
Masterflex® Plastic Ball Valves, Three-way, PVC, Avantor®
Valves Ball Valves
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
  • Maximum pressure: 125 psi (8.6 bar)
  • Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.