Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Supplier: MIDWEST VALVE & FITTING INC
TA-BV-8-8P TA-BV-12P BV-16-16-P
MFLX98514-01EA 1643.2 CAD
MFLX98514-01 MFLX98514-05 MFLX98514-09
Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Valves Ball Valves
PTFE wetted parts allow for maximum chemical resistance.
  • Bidirectional 1/4-turn from OFF to ON
  • PTFE and polypropylene (PP) body construction
  • Perfect for use in harsh environments
These Masterflex® PTFE ball valves are ideal for use in semiconductor or pharmaceutical manufacturing and chemical processing.