Masterflex® Manifold Fittings, Female Luer Locks, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
B14312FMH B14343FMH (10PK) B14314FMH (10PK) B14312FMH (10PK) B14345FMH (10PK)
MFLX30600-42CS 0 CAD
MFLX30600-42 MFLX30600-41 MFLX30600-44 MFLX30600-43 MFLX06464-81
Masterflex® Manifold Fittings, Female Luer Locks, Avantor®
Valves Ball Valves
Regulate flow without any leaking.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.