Masterflex® Large-Bore Stopcock Fittings, Male Luer Lock, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
S3302556 100/PK S3302556 (10PK) B19311LG2 (10PK) B19311LG2 100/PK (MEDEX)
MFLX30600-12CS 0 CAD
MFLX30600-12 MFLX30600-23 MFLX30600-17 MFLX30600-25
Masterflex® Large-Bore Stopcock Fittings, Male Luer Lock, Avantor®
Valves Ball Valves
Enhance fluid flow with a larger bore than standard luer stopcocks.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Large-bore adapters are perfect for handling higher flow volumes in process while using connections similar to luer systems.