Masterflex® Multi-Turn PTFE Needle Valves, Avantor®
Supplier: VWR International
51L 51SL 51SH 51GSH 51TL 51GSL 51TH
MFLX06393-60EA 104.12 CAD
MFLX06393-60 MFLX06393-61 MFLX06393-70 MFLX06393-71 MFLX06393-80 MFLX06393-82 MFLX06393-92 MFLX06393-94 MFLX06393-96 MFLX06393-66 MFLX06393-90 MFLX06393-68
Masterflex® Multi-Turn PTFE Needle Valves, Avantor®
Valves Needle Valves
Easily meter corrosive fluids or gases.
  • Constructed of durable PTFE
Maximum pressure is 75 psi; maximum temperature is 200 °F (93 °C). Wetted parts are body and O-ring made of PTFE and CTFE valve spindle. Choose aluminum (Al) or 316 stainless steel (SS) body. Orifice is 0.125".