Check Valves with Pipe Thread Connections
Supplier: VWR International
MFLX01350-30 MFLX01350-36 MFLX01350-20 MFLX01350-24 MFLX01350-26 MFLX01350-32
MFLX01350-30EA 379.2 CAD
MFLX01350-30 MFLX01350-36 MFLX01350-20 MFLX01350-24 MFLX01350-26 MFLX01350-32
Check Valves with Pipe Thread Connections
Valves Check Valves
Create a secure pipe threaded connection.
  • Choose from PVDF, PVC, or polypropylene (PP) bodies
All valves have Viton® diaphragms.