Masterflex® Transfer Tubing, Gum Rubber, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
11538 120PXA-25' 11542 073PXA-25' 075PXA-25' 118PXA-25' 138PXA-25' 099PXA-25' 122PXA-25' 124PXA-25' 11534 11541 070PXA-25' 140PXA-25' 11544 154PXA-25'
MFLX06448-07EA 96.7 CAD
MFLX06448-07 MFLX06448-17 MFLX06448-06 MFLX06448-05 MFLX06448-04 MFLX06448-03 MFLX06448-13 MFLX06448-02 MFLX06448-23 MFLX06448-12 MFLX06448-11 MFLX06448-09 MFLX06448-08 MFLX06448-21 MFLX06448-10 MFLX06448-20
Masterflex® Transfer Tubing, Gum Rubber, Avantor®
Tubing Transfer Tubing
Keeps shape and cleanliness.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The gum rubber tubing is ideal for general-purpose liquid and gas applications. Manufactured according to GMP, the tubing is nontacky and seamless with a temperature range of -15 to 158 °F (-26 to 70 °C). Can be sterilized with ethylene oxide.