Masterflex® Transfer Tubing, Versilon™ C-544-A IB, Avantor®
Supplier: VWR International
MFLX95637-02 MFLX95637-00 MFLX95637-01 MFLX95637-04 MFLX95637-05 MFLX95637-06 MFLX95637-07
MFLX95637-02EA 1275.06 CAD
MFLX95637-02 MFLX95637-00 MFLX95637-01 MFLX95637-04 MFLX95637-05 MFLX95637-06 MFLX95637-07
Masterflex® Transfer Tubing, Versilon™ C-544-A IB, Avantor®
Tubing Transfer Tubing
No plasticizers.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The Versilon™ C-544-A IB ether-based polyurethane pressure tubing, is ideal for abrasive and viscous slurry transfer, instrumentation control lines, food and cosmetic processing, and coolant recovery systems. Characteristics include exceptional abrasion and tear resistance; excellent resistance to oils, grease and fuels. The transparent tubing features plasticizer-free construction and has a temperature range of -100 to 180 °F (-73.3 to 82.2 °C). It is FDA, NSF 51, and NSF 61 compliant. Not recommended for sterilization.