Masterflex® Transfer Tubing, Metric Polytetrafluoroethylene (PTFE), Avantor®
Supplier: VWR International
MFLX06363-10 MFLX06363-20 MFLX06605-52 MFLX06605-53 MFLX06605-54 MFLX06605-55 MFLX06605-56 MFLX06605-57 MFLX06363-30
MFLX06363-10EA 241.74 CAD
MFLX06363-10 MFLX06363-20 MFLX06605-52 MFLX06605-53 MFLX06605-54 MFLX06605-55 MFLX06605-56 MFLX06605-57 MFLX06363-30
Masterflex® Transfer Tubing, Metric Polytetrafluoroethylene (PTFE), Avantor®
Tubing Transfer Tubing
Withstands extreme temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent.
Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP 23 Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -400 to 500 °F (-240 to 260 °C)
Sterilize: With autoclave, ethylene oxide, or use dry heat.