Masterflex® Transfer Tubing, Peroxide-Cured PTFE, Green, Avantor®
Supplier: VWR International
MFLX96130-42 MFLX96130-44
MFLX96130-42EA 105.07 CAD
MFLX96130-42 MFLX96130-44
Masterflex® Transfer Tubing, Peroxide-Cured PTFE, Green, Avantor®
Tubing Transfer Tubing
Quickly identify lines.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Ideal for dedicated or multiple lines—provides quick and easy line identification.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Opaque.
Certifications: None.
Temperature range: -400 to 500 °F (-240 to 260More Product Information