Masterflex® Transfer Tubing, FEP-Lined Polyethylene, Avantor®
Supplier: VWR International
MFLX06385-02 MFLX06385-03 MFLX06385-06
MFLX06385-04EA 1164.46 CAD
MFLX06385-04 MFLX06385-02 MFLX06385-03 MFLX06385-06
Masterflex® Transfer Tubing, FEP-Lined Polyethylene, Avantor®
Tubing Transfer Tubing
Bendable a low temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Ground water sampling and other sampling and monitoring uses that require the high purity and chemical inertness of FEP.
Characteristics: Chemically inert, non-toxic FEP-liner with a durable polyethylene jacket. Rigid butMore Product Information