Masterflex® Transfer Tubing, Tygon® A-60-F, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
AAL00012-CP AAL00038-CP AAL00019-CP AAL00046-CP AAL00029-CP AAL00053-CP AAL00003-CP AAL00022-CP AAL00007-CP AAL00017-CP
MFLX95710-90EA 976.44 CAD
MFLX95710-90 MFLX95710-00 MFLX95710-40 MFLX95710-30 MFLX95710-20 MFLX95710-10 MFLX95710-80 MFLX95710-70 MFLX95710-60 MFLX95710-50
Masterflex® Transfer Tubing, Tygon® A-60-F, Avantor®
Tubing Transfer Tubing
Stay compliant with food applications.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for the food, dairy, and beverage industries. Characteristics include heat, ozone, and UV light resistant; non-aging; nonoxidizing; superior acid and alkali resistance. Tubing is manufactured according to GMP and has a temperature range of –60 to 275 °F (–51 to 135 °C). Sterilize with autoclave.