Masterflex® Microbore Transfer Tubing, PTFE, Avantor®
Supplier: VWR International
MFLX06417-11 MFLX06417-21 MFLX06417-31 MFLX06417-41 MFLX06417-51 MFLX06417-60
MFLX06417-11CS 0 CAD
MFLX06417-11 MFLX06417-21 MFLX06417-31 MFLX06417-41 MFLX06417-51 MFLX06417-60
Masterflex® Microbore Transfer Tubing, PTFE, Avantor®
Tubing Transfer Tubing
Ideal of extreme temperatures and harsh chemical environments.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Use this translucent tubing in extreme temperatures and in harsh chemical environments. It's chemically inert, has low permeability, and has the lowest friction coefficient of any tubing. TemperatureMore Product Information