Masterflex® Ultramicrobore PTFE Tubing, Avantor®
Supplier: VWR International
MFLX06417-70 MFLX06417-72 MFLX06417-74 MFLX06417-76
MFLX06417-70CS 430.5 CAD
MFLX06417-70 MFLX06417-72 MFLX06417-74 MFLX06417-76
Masterflex® Ultramicrobore PTFE Tubing, Avantor®
Tubing Transfer Tubing
Minimize friction, maximize chemical resistance.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Use this translucent tubing in extreme temperatures and in harsh chemical environments. It's chemically inert, has low permeability, and has the lowest friction coefficient of any tubing. Temperature range is -400More Product Information