Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ 2001, Avantor®
Supplier: VWR International
MFLX06475-15 MFLX06475-24
MFLX06475-15EA 474 CAD
MFLX06475-15 MFLX06475-24
Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ 2001, Avantor®
Tubing Peristaltic Pump Tubing
Compatible with aggressive chemicals.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Chemical, food, beverage, and pharmaceutical applications
Characteristics: Excellent chemical resistance, low extractability, low gas permeability, plasticizer free, translucent
NOTE: Tygon® Chemical pump tubing is recommended for use in Easy-Load® and Easy-Load® II pump heads only.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.