Masterflex® L/S® High-Pressure Pump Tubing, Tygon® A-60-F High Pressure, Avantor®
Supplier: VWR International
MFLX06504-15 MFLX06504-16
MFLX06504-15EA 320.74 CAD
MFLX06504-15 MFLX06504-16
Masterflex® L/S® High-Pressure Pump Tubing, Tygon® A-60-F High Pressure, Avantor®
Tubing Peristaltic Pump Tubing
Designed to pump at higher pressures.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance
Characteristics: Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance, opaque black
Note: L/S® High-Pressure pump tubing is designed for use with the L/S® High-Performance pump head models 77252-62, 77252-72, and 77252-82 only. Maximum drive speed for the L/S® High-Pressure pump tubing is 100 rpm.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.