Masterflex® I/P® Spooled Pump Tubing, Peroxide-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96402-73 MFLX96402-82 MFLX96402-26
MFLX96402-73EA 2543.8 CAD
MFLX96402-73 MFLX96402-82 MFLX96402-26
Masterflex® I/P® Spooled Pump Tubing, Peroxide-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Reduce the risk of particulate entrapment with smooth inner surface.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Economical tubing with a longer tubing life