Masterflex® L/S® Spooled Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96423-14 MFLX96423-15 MFLX96423-16 MFLX96423-17 MFLX96423-18 MFLX96423-24 MFLX96423-25
MFLX96423-14EA 3713 CAD
MFLX96423-14 MFLX96423-15 MFLX96423-16 MFLX96423-17 MFLX96423-18 MFLX96423-24 MFLX96423-25
Masterflex® L/S® Spooled Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Ultra-smooth inner surface minimizes particle entrapment.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications
Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.