Masterflex® I/P® High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96421-70 MFLX96421-88 MFLX96421-89
MFLX96421-70CS 1053 CAD
MFLX96421-70 MFLX96421-88 MFLX96421-89
Masterflex® I/P® High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Smooth inner surface designed to reduce risk of particle entrapment and contamination.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
Choose our BioPharm SiliconeMore Product Information