Masterflex® L/S® Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96420-13 MFLX96420-14 MFLX96420-16 MFLX96420-17 MFLX96420-18 MFLX96420-25
MFLX96420-13EA 235.42 CAD
MFLX96420-13 MFLX96420-14 MFLX96420-16 MFLX96420-17 MFLX96420-18 MFLX96420-25
Masterflex® L/S® Precision Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Ultra-smooth inner surface minimizes particle entrapment.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications
Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.