Masterflex® L/S® Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Supplier: VWR International
AAG00122-CP AAG00151-CP AAG00192-CP AAG00248-CP AAG00259-CP AAG00863-CP
MFLX06401-13EA 133.67 CAD
MFLX06401-13 MFLX06401-14 MFLX06401-16 MFLX06401-17 MFLX06401-18 MFLX06401-25
Masterflex® L/S® Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Tubing Peristaltic Pump Tubing
Ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Fuels and industrial lubricants—gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants
Characteristics: Resists most hydrocarbons, embrittlement, and swelling; ozone- and UV-resistant, low extractability, thermoplastic, PVC-based material with plasticizer, firm (stiff) material, translucent yellow
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.