Masterflex® L/S® Precision Pump Tubing, Versilon™ A-60-N, Avantor®
Supplier: VWR International
AFL00122-CP AFL00151-CP AFL00192-CP AAN00248-CP AAN00259-CP AFL00863-CP
MFLX06404-13CS 132.56 CAD
MFLX06404-13 MFLX06404-14 MFLX06404-16 MFLX06404-17 MFLX06404-18 MFLX06404-25
Masterflex® L/S® Precision Pump Tubing, Versilon™ A-60-N, Avantor®
Tubing Peristaltic Pump Tubing
Well-suited for vacuum/pressure applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance
Characteristics: Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance, opaque black
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.