Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96410-13 MFLX96410-14 MFLX96410-16 MFLX96410-17 MFLX96410-18 MFLX96410-25
MFLX96410-13CS 145.36 CAD
MFLX96410-13 MFLX96410-14 MFLX96410-16 MFLX96410-17 MFLX96410-18 MFLX96410-25
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.