Masterflex® B/T® PerfectPosition® Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96510-87 MFLX96510-91 MFLX96510-92
MFLX96510-87EA 996.98 CAD
MFLX96510-87 MFLX96510-91 MFLX96510-92
Masterflex® B/T® PerfectPosition® Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Ensure optimal performance from your Masterflex® B/T series pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and
More Product Information