Masterflex® I/P® High-Performance Precision Pump Tubing, C-Flex®, Avantor®
Supplier: VWR International
190-925-001 190-724-001 190-926-001
MFLX06424-70EA 281.24 CAD
MFLX06424-70 MFLX06424-88 MFLX06424-89
Masterflex® I/P® High-Performance Precision Pump Tubing, C-Flex®, Avantor®
Tubing Peristaltic Pump Tubing
Maintain flexibility at extreme temperatures.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Ultra smooth inner surface results in fewer extractables
Masterflex® C-Flex® High- Performance Precision pump tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic — protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.