Masterflex® I/P® Precision Pump Tubing, Peroxide-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96400-26 MFLX96400-73 MFLX96400-82
MFLX96400-26CS 357 CAD
MFLX96400-26 MFLX96400-73 MFLX96400-82
Masterflex® I/P® Precision Pump Tubing, Peroxide-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Reduce the risk of particulate entrapment with smooth inner surface.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Economical tubing with a longer tubing life
Masterflex® peroxide-cured silicone High-Performance Precision pump tubing features greater physical compression capability with a longer pumping life compared to platinum-cured silicone tubing. Translucent tubing is made with no additives or plasticizers and makes for easy flow monitoring. Ideal for general-purpose, biotech, and food and beverage applications, this tubing complies with USP Class VI, FDA, and 3-A standards.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.