Masterflex® Liquid Detector Pad Cables, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
77095-02 77095-00
MFLX77095-00EA 265.44 CAD
MFLX77095-00 MFLX77095-02
Masterflex® Liquid Detector Pad Cables, Avantor®
Cables
Make the most of your Liqui-Sense leak detector with suitable replacement parts.
  • Compatible with Masterflex® Liqui-Sense controllers
This replacement cable features a 6-pin cable with RJ-12 connections and has a 3.5 foot cable length.