Masterflex® Solenoid Valve Connection Cable, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
CBL-SMV
MFLX98651-50EA 83.9 CAD
MFLX98651-50
Masterflex® Solenoid Valve Connection Cable, Avantor®
Cables
Easy connect to your solenoid valve.
  • Optimize performance of your Masterflex® Solenoid Valves
"D" connector connection cable with stripped wire ends for Aalborg stepping motor proportioning valves. This cable is 3-ft (0.9m) long.