Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
Supplier: VWR International
MFLX32774-60 MFLX32774-62 MFLX32774-64 MFLX32774-66 MFLX32774-68
MFLX32774-60EA 323.9 CAD
MFLX32774-60 MFLX32774-62 MFLX32774-64 MFLX32774-66 MFLX32774-68
Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
Flow Switches
Connects quickly to threaded plastic piping.
  • Chemically resistant — wetted materials stainless steel and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • NSF and FDA approved
These normally open switches have 3/4" NPT(F) ports for a quick connection to threaded plastic piping. Polypropylene and stainless steel wetted parts offer excellent chemical resistance. The materials are also NSF approved and FDA compliant; approved for potable water treatment applications such as chlorinators, purifiers, and heaters. Switches include 24" lead wires.