Masterflex® Tank Dispensing Spray Guns, Avantor®
Supplier: DYTEC CORPORATION
THG-DN6P-TFEV THG-LD6P-TFEV
MFLX98515-01EA 323.9 CAD
MFLX98515-01 MFLX98515-03 MFLX98515-05 MFLX98515-07 MFLX98515-09
Masterflex® Tank Dispensing Spray Guns, Avantor®
Spray Guns
No splashing.
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.