Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, PTFE, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
F-MH0604 F-MH0403 F-MH0808 F-MH0303 F-MH0203 F-MH0302 F-MH0102 F-MH0304 F-MH0402 F-MH0404 F-MH0202 F-MH0201 F-MH0806 F-MH0101
MFLX34030-81EA 246.48 CAD
MFLX34030-81 MFLX34030-80 MFLX34030-79 MFLX34030-83 MFLX34030-82 MFLX34030-74 MFLX34030-84 MFLX34030-73 MFLX34030-87 MFLX34030-76 MFLX34030-75 MFLX34030-86 MFLX34030-78 MFLX34030-77
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, PTFE, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.