Masterflex® Reducer Fittings, Hose Barb, Straight (Polypropylene), Avantor®
Supplier: VWR International
MFLX30616-51 MFLX30616-63 MFLX30616-67 MFLX30616-71 MFLX30616-75 MFLX30705-12 MFLX40610-07 MFLX40610-08 MFLX40610-09 MFLX40610-10 MFLX40610-11 MFLX40610-12 MFLX40610-13 MFLX40610-14 MFLX40610-15 MFLX40610-16 MFLX40610-17 MFLX40610-18 MFLX40610-19 MFLX40610-20 MFLX40610-22 MFLX40616-43 MFLX40616-47 MFLX40622-19 MFLX40622-26 MFLX40622-32 MFLX40626-26 MFLX40626-32 MFLX40640-08 MFLX40640-09 MFLX40640-10 MFLX40640-12 MFLX40640-13 MFLX40640-18 MFLX40640-20 MFLX40920-23 MFLX41518-34 MFLX41518-36 MFLX50621-95 MFLX50622-21 MFLX50622-23 MFLX50626-21 MFLX50626-23 MFLX40610-21 MFLX40640-11 MFLX40640-15 MFLX40640-16 MFLX40640-17 MFLX40640-21 MFLX50625-95 MFLX30705-11
MFLX30616-51EA 46.45 CAD
MFLX30616-51 MFLX30616-63 MFLX30616-67 MFLX30616-71 MFLX30616-75 MFLX30705-12 MFLX40610-07 MFLX40610-08 MFLX40610-09 MFLX40610-10 MFLX40610-11 MFLX40610-12 MFLX40610-13 MFLX40610-14 MFLX40610-15 MFLX40610-16 MFLX40610-17 MFLX40610-18 MFLX40610-19 MFLX40610-20 MFLX40610-22 MFLX40616-43 MFLX40616-47 MFLX40622-19 MFLX40622-26 MFLX40622-32 MFLX40626-26 MFLX40626-32 MFLX40640-08 MFLX40640-09 MFLX40640-10 MFLX40640-12 MFLX40640-13 MFLX40640-18 MFLX40640-20 MFLX40920-23 MFLX41518-34 MFLX41518-36 MFLX50621-95 MFLX50622-21 MFLX50622-23 MFLX50626-21 MFLX50626-23 MFLX40610-21 MFLX40640-11 MFLX40640-15 MFLX40640-16 MFLX40640-17 MFLX40640-21 MFLX50625-95 MFLX30705-11
Masterflex® Reducer Fittings, Hose Barb, Straight (Polypropylene), Avantor®
Fittings and Connectors
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range 9 to 180 °F (-13 to 66 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.